Learn More
Low Risk First Aid Kit
Administer to those requiring emergency care with the Low Risk First Aid Kit, available for non-critical nor hazardous emergencies. Kits contain the basic elements necessary to provide emergency care in the event is is necessary.
Brand: Safety First K20T/
13.01 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 50
Additional Details : Weight : 1.00000kg
Description
- First Aid Guidance Leaflet
- Wash proof Plasters
- Sterile Eye Pads With Bandage
- Triangular Bandages
- Safety Pins
- First Aid Dressings
- Moist Wipes
- Disposable Gloves
Specifications
First Aid Kit | |
Medium | |
295 x 225 x 117 mm |
1 Ea. | |
11 to 20 Persons |