Learn More
Fisher BioReagents™ Adenosine 5'-triphosphate Disodium Salt Trihydrate
Brand: Fisher BioReagents™ BP413-25
162.84 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 32
Additional Details : CAS Number : 51963-61-2 Weight : 0.07500kg
Description
Disodium Salt ATP
- C10H14N5O13P3Na2·3H2O; F.W. 605.24
- CAS: 51963-61-2
- Molar Extinction Coefficient (259nm, phosphate buffer, pH 7.0); >15,000
- Solubility (5% in H2O, 25°C): clear and haze-free
- Lead: ≤0.002%
Suitable for use in many molecular biology applications
Specifications
Solid | |
605.24 | |
25 g | |
Molecular biology |
7 | |
<0°C | |
White |