missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ ChromoMax™ IPTG/X-Gal Solution

Accelerate your blue/white colony screening

£30.23 - £48.36

Specifications

Content And Storage -20°C
Additive IPTG/X-Gal Solution
Form Liquid
For Use With (Application) All commercially available competent cells that utilize lacZ blue/white screening
Packaging Type Bottle
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10133443
View Documents
Fisher BioReagents™
BP4200-1
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10225163
View Documents
Fisher BioReagents™
BP4200-10
10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Ready-to-apply ChromoMax IPTG/X-Gal solution is a proprietary formulation that allows you to skip the cumbersome preparation process that traditionally requires tedious weighing and dissolving of fine powder in toxic solvents such as DMSO or DMF.

  • Premixed IPTG/X-Gal Solution
  • No weighing or dissolving of components
  • No DMSO or DMF required
  • Saves time and money
  • Dependable results

  • Can be safely handled outside of a fume hood
  • ChromoMax Solution does not freeze even when stored at -20°C, allowing for immediate use without thawing
  • Dries quickly on the plate, further accelerating the screening protocol
  • Provides fast color development and does not require additional 4°C incubation to enhance color
  • Clear colorless to yellow solution

ChromoMax/ITPTG/X-Gal Solution is suitable for use with all commercially available competent cells that utilize lacZ blue/white screening. Tested using Fisher BioReagents transformed TransMax Competent Cells resulting in clearly differentiated blue and white colonies.

Order Info

The 50mL volume is conveniently packaged in five 10mL bottles, preventing accidental contamination of large volume stock.

Specifications

Specifications

-20°C
Liquid
Bottle
IPTG/X-Gal Solution
All commercially available competent cells that utilize lacZ blue/white screening
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ChromoMax

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H360D-May damage the unborn child.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ ChromoMax™ IPTG/X-Gal Solution

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.