Learn More
Fisher BioReagents™ EZ-Run™ Prestained Rec Protein Ladder, Fisher BioReagents
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
£201.00 - £452.00
Specifications
Color | Blue |
---|---|
Product Type | Protein Molecular Weight Marker/Ladder |
Form | Liquid |
Formulation | Supplied in gel-loading buffer |
For Use With (Application) | Monitoring protein separation during SDS-PAGE |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
10638393
|
Fisher BioReagents™
BP3603-500 |
500 μL, 100 Loadings | 100 Loadings |
£201.00
Each |
In Stock
Log in to see stock. |
||||
10785674
|
Fisher BioReagents™
BP3603-1 |
2 x 500 μL, 200 Loadings | 200 Loadings |
£452.00
Pack of 2 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
Description
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
- Ladder is ideal for monitoring protein separation during SDS-PAGE, verifying Western blot transfer efficiency and approximating the molecular weight of blotted proteins
- Supplied in gel loading buffer
The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.
Specifications
Blue | |
Liquid | |
Monitoring protein separation during SDS-PAGE | |
−20°C |
Protein Molecular Weight Marker/Ladder | |
Supplied in gel-loading buffer | |
10-170 kDa |
This product is manufactured under the license for Strep-tag™ technology covered by U.S. Patent No. 5,506,121; U.K. Patent No. 2272698; and French Patent No. 93 13 066.