Learn More
Fisher BioReagents™ EZ-Run™ Prestained Protein Marker
Ideal for monitoring protein separation during electrophoresis without staining and for verifying electroblotting of proteins onto membranes
£147.30
Specifications
Product Type | Protein Molecular Weight Marker/Ladder |
---|---|
Form | Liquid |
Formulation | Supplied in gel-loading buffer |
For Use With (Application) | Monitoring protein separation during electrophoresis |
Kit Contents | <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
10408923
|
Fisher BioReagents™
BP3601-1 |
2 x 500 μL, 200 Loadings | 200 Loadings | N/A | |||||
Description
- Comprised of six purified proteins covalently coupled to a blue chromophore
- Optimized to produce well-defined blue bands after SDS-PAGE
- Supplied in gel loading buffer
- 20 to 118kDa
The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.
The Prestained Protein Marker is designed for monitoring protein separation during electrophoresis without staining. It is also used to verify electroblotting of proteins onto membranes.
Specifications
Protein Molecular Weight Marker/Ladder | |
Supplied in gel-loading buffer | |
<51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet | |
−20°C |
Liquid | |
Monitoring protein separation during electrophoresis | |
20 to 118 kd |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.