missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ exACTGene™ Control Primers

Product Code. 11428293
Click to view available options
Product Type:
Control Primer No. 1
Control Primer No. 2
Control Primer No. 3
Quantity:
1 Kit
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 11428293

Brand: Fisher BioReagents™ BP6117

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimized for routine PCR applications

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 μm
Content And Storage -20°C
For Use With (Equipment) Fisher BioReagents™ exACTGene™ PCR kit
Product Type Control Primer No. 2
Quantity 1 Kit
Volume 100μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.