missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Broth, Lennox (Granulated)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
£70.77 - £276.28
Specifications
Form | Granulated Powder |
---|---|
Media Type | LB Broth, Lennox |
Formulation | Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11315992
|
Fisher BioReagents™
BP9722-2 |
Poly Bottle | 2000 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11305992
|
Fisher BioReagents™
BP9722-500 |
Poly Bottle | 500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Directions for Media Preparation: Add 20g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Provides a nutrient-rich environment to enable faster growth of E. coli
- Format: Granulated powder
Specifications
Granulated Powder | |
LB Broth, Lennox | |
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L |