missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Lysozyme, Egg White (White Cryst. Powder)

£31.30 - £314.57

Specifications

Color White
For Use With (Application) Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
Content And Storage <0°C
Source Egg white
Product Type Lysozyme
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10536394
View Documents Promotion Details
Fisher BioReagents™
BP535-1
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10535054
View Documents Promotion Details
Fisher BioReagents™
BP535-5
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10421545
View Documents Promotion Details
Fisher BioReagents™
BP535-10
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10421735
View Documents Promotion Details
Fisher BioReagents™
BP535-25
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Application: This enzyme is used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
  • Enzyme Activity: ≈20,000U/mg dry weight
  • Solubility: Readily soluble at 5mg/mL in 0.1M KH2PO4 to give colorless solution
Specifications

Specifications

White
<0°C
Lysozyme
Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
Egg white
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lysozyme, egg white

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Product Certifications