missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media Additives: Yeast Extract
Additive for preparing bacterial nutrient media
£31.55 - £272.00
Specifications
Form | Dehydrated |
---|---|
Grade | Molecular genetics |
Media Type | Yeast Extract |
Product Type | Microbiology Media |
Packaging Type | Poly Bottle |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10255153
|
Fisher BioReagents™
BP1422-100 |
100 g |
£31.55
100g |
In Stock
Log in to see stock. |
|||||
10225203
|
Fisher BioReagents™
BP1422-500 |
500 g |
£109.00
500g |
In Stock
Log in to see stock. |
|||||
10215203
|
Fisher BioReagents™
BP1422-2 |
2000 g |
£272.00
2kg |
In Stock
Log in to see stock. |
|||||
Description
Water soluble portion of autolyzed fresh yeast used in bacteriological culture mediaSpecifications
Dehydrated | |
Yeast Extract | |
Poly Bottle |
Molecular genetics | |
Microbiology Media |