missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder)
Click to view available options
Quantity:
500 g
2000 g
Unit Size:
2kg
500g
Description
- Directions for Media Preparation: Suspend 40g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
- Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
Specifications
Specifications
| Description | Bacterial Culture |
| Form | Powder |
| Grade | Molecular genetics |
| Media Type | LB Agar |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L |
| For Use With (Application) | For preparing bacterial nutrient media |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder) >
Spot an opportunity for improvement?Share a Content Correction