missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Brand: Fisher BioReagents™ BP1425-2
This item is not returnable.
View return policy
Description
- Directions for Media Preparation: Suspend 40g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
- Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain

Specifications
Dehydrated Powder | |
LB Agar | |
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L | |
Poly Bottle |
Molecular genetics | |
Agar | |
Preparing bacterial nutrient media | |
2000 g |
Safety and Handling
Product Identifier
- LB Agar, Miller
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder) > Agar Miller; 2kg
Spot an opportunity for improvement?Share a Content Correction