missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media: LB Broth, Miller

Used for growing molecular biology strains of E. coli-containing recombinant plasmids

£56.10 - £179.85

Specifications

Content And Storage 4°C
Form Powder
Grade Molecular genetics
Media Type LB Broth
Product Type Microbiology Media
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10638013
View Documents
Fisher BioReagents™
BP1426-2
2000 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10113293
View Documents
Fisher BioReagents™
BP1426-500
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Dehydrated.

Specifications

Specifications

4°C
Molecular genetics
Microbiology Media
For preparing bacterial nutrient media
Powder
LB Broth
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Broth Miller

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.