missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Peptone (Granulated)
£101.60 - £322.42
Specifications
Form | Granulated Powder |
---|---|
Grade | Molecular genetics |
Media Type | Peptone |
Product Type | Peptone |
Formulation | Total Nitrogen <10%, Amino Nitrogen <3.5% |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11395982
|
Fisher BioReagents™
BP9725-2 |
Poly Bottle | 2 kg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11375982
|
Fisher BioReagents™
BP9725-500 |
Poly Bottle | 500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue
- pH (at 25°C, 2% Solution): 6.5 to 7.5
- CAS: 73049-73-7
Specifications
Granulated Powder | |
Peptone | |
Total Nitrogen <10%, Amino Nitrogen <3.5% |
Molecular genetics | |
Peptone |