missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Restriction Enzymes - Hind III (Source: Haemophilus influenzae Rd)
£17.30
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428333
|
Fisher BioReagents™
BP3372-1 |
5000 U | N/A | ||||||
Description
- Application: Hind III restriction endonuclease digests dsDNA with sequence indicated below
- Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 250mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
- Qualified for Blue/White Cloning Assays
- Storage: -20°C
Specifications
10 U/μL | |
A.AGCTT | |
HindIII |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title