Learn More
Fisher BioReagents™ SurePrep™ Urine Bacterial RNA Purification Kit
Product Code. 10582744
Click to view available options
Quantity:
50 Preps
Unit Size:
Pack of 50
Product Code. 10582744
Brand: Fisher BioReagents™ BP280450
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Lysis solution (40mL); Wash solution (22mL); RNA elution buffer (6mL); Spin columns (50); Collection tubes (50); Elution tubes (1.7mL) (50); Product insert (1)
Designed for the rapid preparation of bacterial RNA from human or animal urine samples
  • Fast and easy processing using rapid spin-column format
  • Isolate total RNA, from large mRNA down to microRNA (miRNA)
  • No phenol or chloroform extractions
  • Isolate RNA from both Gram negative and Gram positive bacteria, including E. coli, Proteus spp., Klebsiella spp., Enterobacter spp., Serratia spp., Pseudomonas spp., Clostridial ssp. and Leptospirosis spp., as well as Chlamydia trachomatis and Neisseria gonorrhoeae
Purified bacterial RNA suitable for real-time PCR, reverse transcription PCR, Northern blotting, RNase protection, primer extension, and expression array assays.
TRUSTED_SUSTAINABILITY
Specifications
Format Spin Column
For Use With (Application) Purified bacterial RNA suitable for RT-PCR, reverse transcription PCR, northern blotting, RNase protection, primer extension and expression array assays
Includes Lysis solution 40mL, wash solution 22mL, RNA Elution Buffer 6mL, spin columns 50, collection tubes 50, 50 elution tubes 1.7mL, product insert 1
Quantity 50 Preps
Yield ≈1μg RNA/1 x 105 cells