missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Tryptone (Granulated)
An enzymatic digest of casein, a milk protein
Brand: Fisher BioReagents™ BP9726-2
This item is not returnable.
View return policy
Description
- pH (at 25°C, 2% Solution): 6.5 to 7.3
Specifications
Granulated Powder | |
Microbiology Media | |
Poly Bottle |
Tryptone | |
Total Nitrogen ≥10%, Amino Nitrogen ≥3.5% | |
2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Tryptone (Granulated) > 2kg
Spot an opportunity for improvement?Share a Content Correction