missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Automatic Temperature Compensation Probes - Mercury - Free

For accumet™ benchtop meters.

£223.00 - £305.00

Specifications

Type ATC
Length (Metric) Cable 0.9 m
View More Specs

Products 3
Product Code Brand Connections Material For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Connections Material For Use With (Equipment) Price Quantity & Availability  
11530234
View Documents Promotion Details
Fisherbrand™
01X306701
0.25 in. Phone Stainless Steel Accumet benchtop and portable meter
£305.00
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
 
10236064
View Documents Promotion Details
Fisherbrand™
01X306501
Mini-Phone 316 Stainless Steel accumet AB, AR, XL, Basic, benchtop meters
£223.00
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
 
11540234
View Documents
Fisherbrand™
01X306601
Mini-Phone Stainless Steel Accumet model AP63, AP62, AP61 N/A N/A N/A
Description

Description

For use with accumet Meters

  • Signal solution temperature to meter for automatic compensation
  • Permit temperature readout with meters having this feature
  • Probes for accumet AP Series portable meters have waterproof connector

Specifications

Specifications

ATC
0.9 m
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ accumet™ Automatic Temperature Compensation Probes - Mercury - Free

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.