Learn More
Fisherbrand™ accumet™ Conductivity Calibration Kit
Precision calibration resistor kit for accumet conductivity meters.
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Includes:
- Six precision resistors (nominal values 10ohm, 100ohm, 1kohm, 10kohm, 100kohm, and 1Mohm)
- Connectors for accumet XL and AB conductivity meters
- Foam padded case
Description
Precision calibration resistor kit for accumet conductivity meters.
- Supplied with a factory test certificate
Specifications
Conductivity Calibration Kit |