missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Glass Body Calomel Reference Electrodes
£98.43 - £297.68
Specifications
Connector Types | Pin |
---|---|
Material | Glass |
pH Range | 0 to 14 |
Temperature (Metric) Operating | -5°C to +80°C |
Filling Solution Type | Mercury |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | For Use With (Application) | Diameter (Metric) Cap | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Diameter (Metric) Cap | Length (English) | Price | Quantity & Availability | |||
11523975
|
Fisherbrand™
93X218938 |
Biomedical, Tris | 1.6 cm | 5.75 in. | N/A | ||||
11533975
|
Fisherbrand™
93X218898 |
Biomedical, Tris | 1.6 cm | 5.75 in. | N/A | ||||
Description
- Standard-size glass body
- Contains mercury
- -5° to +80°C
- 0 to 14pH range
Specifications
Pin | |
0 to 14 | |
Mercury | |
Calomel | |
5.75 in. | |
14.6 cm | |
0.37 in. | |
1.6 cm |
Glass | |
-5°C to +80°C | |
Hg/Hg2Cl2 (Calomel) | |
23°F to 176°F | |
30 in. | |
76.2 cm | |
1 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title