missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Liquid-Filled Mercury-Free pH/ATC Epoxy Body Combination Electrodes, w/BNC and Mini-phone ATC connectors
Shop All Fisherbrand Products
Click to view available options
Temperature (Metric) Operating:
Unit Size:
Each
Description
Includes
Electrode, 60mL refill solution, electrode storage bottle
Requires
Requires filling solution
TRIS compatible, rugged, and convenient
- Rugged standard-size epoxy body
- Double-junction design prevents reference contamination; compatible with TRIS applications; mercury free
- Liquid filled design provides quick response
- Porous ceramic junction, Ag/AgCl reference
- Built-in ATC probe
- Rugged epoxy body for use in field or plant
- Ideal for handheld use with aqueous samples
- Double-junction design eliminates measurement problems and clogging in research with TRIS buffers
Order Info
Endcap seal; keeps membrane moist and ready
Specifications
Specifications
| Color | Purple |
| Connector Types | BNC/Mini-Phone ATC |
| Junction | Porous Ceramic Double |
| Material | Epoxy |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | pH/ATC Combination Titration Electrode |
| Filling Solution Type | Saturated KCl Electrolyte (SP138-500) |
| For Use With (Application) | General laboratory and life science applications |
| For Use With (Equipment) | Accumet Excel (XL) series, Research (AR) and Basic (AB15 and AB15 Plus) benchtop meter |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ accumet™ Liquid-Filled Mercury-Free pH/ATC Epoxy Body Combination Electrodes, w/BNC and Mini-phone ATC connectors >
Spot an opportunity for improvement?Share a Content Correction





