missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Mercury-free Conductivity Probes: Two Cell

For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters

Brand:  Fisherbrand™ 93X219028

 View more versions of this product

Product Code. 11510174

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Requires : Separate ATC probe for temperature-compensated readings.
This item is not returnable. View return policy

Description

Description

For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters

Immersion-type cell's unique compact size is the same as a typical pH electrode—allowing use in standard electrode holder.

  • Two-cell types with glass or epoxy bodies
  • Dual pin connectors
  • Cell constants of 0.1, 1.0, or 10.0cm-1

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 cm-1
AR60, AR50, AR20, AB30 benchtop and portable meter
Epoxy
Immersed in deionized water
2-pin
Conductivity Probe
10 to 2000 μS/cm
2 Cell Conductivity
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers