missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Brand:  Fisherbrand™ 93X218909

147.92 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11590104

  • £259.00 / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Zero to 14pH range
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BNC
Glass
-5°C to +110°C
Saturated KCl with AgCl. KCl
Ag/AgCl
-5°C to +110°C
1 Year
Single
0 to 14
Combination pH electrode, Ag/AgCl
Small samples
23°F to 230°F
Micro
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers