missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Solid State Combination ISEs, Mercury-Free: Silver/Sulfide

For bromide, chloride, copper, cyanide, fluoride, lead and silver/sulfide determinations

Brand:  Fisherbrand™ 93X561501

461.23 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 11590144

  • £669.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For bromide, chloride, copper, cyanide, fluoride, lead and silver/sulfide determinations

Features

  • Low drift, fast response, high selectivity ratio, good thermal stability and wide operating range
  • Continuous electrical shielding and insulation of internal elements, cable and connector
  • Extremely stable, reproducible readings with a minimum of electrical noise
  • Rapid response time that lowers cost per test
  • Individually tested, serialized to meet GLP requirements and backed by the Fisher Scientific Technical Applications staff

 

Combination models

  • Feature a homogenous sensing membrane for wide application, rapid response, minimum sample retention, improved precision, and lower detection limits
  • In glass or epoxy body incorporating sensing and reference cells in a single probe

Silver/Sulfide Combination ISE
  • Determines silver and sulfide in water and pulping liquors
  • Glass body

 

Cyanide solutions are highly toxic; use a pipet bulb when preparing them. Acidic cyanide solutions produce hydrogen cyanide; procedures with these must be done in a fume hood. Measurements above 10-3M should be done infrequently, as the cyanide ion slowly erodes the sensing membrane.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Used to determine silver and sulfide in water and pulping liquors
0.5 in.
13-620-834
108mm
0.01 to 107900ppm Ag+, 0.003 to 32100ppm S2-
2 to 12
Silver/Sulfide Combination
0°C to 80°C
BNC
13mm
4.25 in.
Glass Body
32° to 176°F
Hg2+
0 to 80°C
Combination ISE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers