missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Waterproof AP71 pH/mV/Temperature Meter Kit
Advanced pH meters in waterproof, dustproof housing
Brand: Fisherbrand™ 01X245207
Includes: Meter Kit package includes meter, pH/ATC electrode, pH calibration buffers, sample bottle, and hard plastic carrying case.
Description
- Ergonomic design fits comfortably in the hand
- Electrode bracket positions electrode for one-handed operation
- Advanced set-up mode lets user customize meter parameters and check electrode slope and offset to assure electrode accuracy
- Read pH, absolute mV or relative mV and temperature
- Accuracy of ±0.01pH and ±0.02mV
- Manual or automatic temperature control
- Five-point push-button pH calibration and automatic buffer recognition (pH 1.68, 4.01, 7.01, 10.01, 12.45)
- HOLD function, Ready indicator and automatic shut-off function
- Read pH, mV or relative mV and temperature in °C
- Memory allows storage and recall of up to 16 readings with corresponding temperatures
Certifications
ISO 9001:2000, CE
Services
Warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
Meter, pH/ATC electrode, pH calibration buffers, sample bottle, hard Plastic carrying case | |
±0.2 from 0 to ±400, ±2 beyond 200 | |
pH, mV, Temperature | |
0.001 | |
0.1 from 0 to ±400, 1 beyondmV | |
0°C to 100°C | |
LCD | |
IP-67 | |
1.0 lb. | |
Multiple |
±0.5°C | |
±0.01 | |
-2 to 16 | |
0 to ±2000 | |
5 | |
0.1°C | |
ISO 9001 2000, CE | |
0.5 kg | |
3 Years | |
Automatic or Manual |