Learn More
Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Brand:  Fisherbrand™ 37020-9014
 View more versions of this product
Product Code. 15192987
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes:
  • A 9/16 in. dia. x 10 in. L support rod
  • Four ft. air hose with double fittings for connection to air supply
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Order Info
Shaft, propeller, stand and clamp available separately
Specifications
Specifications
3000 rpm
0.5 in.
1.5
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
633
10 lb.
50 L
1.27 cm
Safely mixing all types of solvents, lacquers and volatile chemicals
40 to 100 psi
30,000 mPas
4.6 kg