Learn More
Fisherbrand™ Computer Data Acquisition System for Digital Barometer
Keep track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more. Computer Data Acquisition System for Digital Barometer is for use with Fisherbrand™ Traceable™ Digital Barometer.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Keep track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more. Computer Data Acquisition System for Digital Barometer is for use with Fisherbrand™ Traceable™ Digital Barometer.
- Allows up to eight unique Traceable™ meters to simultaneously capture data and send it to directly to email to any smartphone or computer
- Permanently capture readings; keep track of results
- Readings are saved to a file that can be viewed, printed as-is, or merged in a report or spreadsheet format
- Data continues to be collected in background even when user works on other software programs
- 24/7 audible/visual alarm reminds user to check on data
Specifications
Data Acquisition Software | |
Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more | |
6 ft. |
CD, 6 ft. Cable | |
Fisherbrand™ Traceable™ Digital Barometer |