Learn More
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Easy-to-read handheld Traceable™ Digital Thermometer.
Brand:  Fisherbrand™ 90029-10
 View more versions of this product
Product Code. 11749715
  • £43.00 / Each
Explore more special offers
Description
Description
Easy-to-read handheld Traceable™ Digital Thermometer.
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Multi-point calibration on individually-numbered Traceable Certificate assures accuracy from ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA; indicates traceability to NIST.
Specifications
Specifications
DNV, ISO 17025, A2LA
3.56 mm
8 in.
20.32 cm
1 oz.
9.25 in.
280°C
-50°C
-50°C to +280°C
±1°C
0.14 in.
Traceable certificate, Stainless steel probe
235 mm
0.1°C
20 g
536°F
-58°F
-58°F to +536°F
Cuvets, soil, incubators, petroleum products and foods, wet labs, wash down areas and field use
Digital LCD
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products