missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Filling Solutions for accumet™ Double Junction Reference Electrodes

For use with accumet™ Double Junction Reference Electrodes.

Brand:  Fisherbrand™ 01Y562601

59.58 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 12972920

  • £80.40 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with accumet™ Double Junction Reference Electrodes.

Five 60mL bottles

TRUSTED_SUSTAINABILITY
Specifications

Specifications

accumet Double Junction Reference Electrode Outer Chamber
Filling Solution
Five 60mL bottles
Double
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.