missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ 4100 H21P Open Heated PPO Bath Circulators

Open heated baths are made from very rigid polyphenyleneoxide (PPO) and are thermally resistant up to +100°C, making them an excellent alternative to stainless steel.

Brand:  Fisherbrand™ 211116100

1090.71 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11396661

  • £1445.00 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These economical open-bath circulators use water as the heat transfer liquid. Baths have integrated grips and supports and are fitted with bridge plates for placement of the control head.

 

  • Heating capacity up to 1000 watts
  • With 21-liter bath
  • The most economical choice for a heating bath
  • Temperature range: Ambient to +100°C
  • Note: Does not include a lid or cover (sold separately)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4.76 to 5.94 gal.
17
Ambient +10° to 100°C
Does not include a lid or cover (sold separately)
4100 H21P
13.7 x 14.1 x 25.3 in. (34.8 x 35.8 x 64.2 cm)
34.8 cm
35.8 cm
64.2 cm
230 V 50 Hz
50 Hz
18 to 22.5 L
Force
Isotemp™ Open Heated Bath Circulator
17 L/min.; 310 mbar
±0.05°C
13.7 in.
14.1 in.
25.3 in.
7.7 kg
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.