missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Bath Circulator Accessories: Plumbing Packages

Plumbing kits for use with bath circulators and chillers

£257.67 - £284.79

Specifications

Temperature Range -25°C to +100°C
View More Specs

Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11637998
View Documents
Fisher Scientific™
1600147
Plumbing Kit, Insulated
£284.79
Each
In Stock
to see stock.
Add to basket
 
11697988
View Documents
Fisher Scientific™
1600146
Plumbing Kit, Uninsulated
£257.67
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
Description

Description

Plumbing kits for use with bath circulators and chillers

  • Plumbing kit
  • Material: Viton
Specifications

Specifications

-25°C to +100°C
Videos
SDS
Documents

Documents

Product Certifications