Learn More
Thermo Scientific™ Thermo Scientifi™ Chiller External Pressure Reducers
External pressure reducers
Brand:  Thermo Scientific™ 196000000000
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11977514
  • £269.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
External pressure reducers for ThermoChill Recirculating chillers
  • 1/2 in. external pressure reducer
  • Adjustable to safely reduce or limit system pressure and flow
Specifications
Specifications
1/2 in. External Pressure Reducer for ThermoChill I & II
ThermoChill Recirculating Chiller I & II
External Pressure Reducer (EPR)
1/2 in.