Learn More
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.
£502.00 - £615.00
Specifications
Certifications/Compliance | CE, cULus, RoHS |
---|---|
Controller Type | PID |
Material | Powder-coated Steel |
Relative Humidity | <80% |
Temperature Range (Metric) | Ambient +5° to 130°C |
Requires: Unit must be ordered with block(s)
Product Code | Brand | No. of Places | Electrical Requirements | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Places | Electrical Requirements | Price | Quantity & Availability | |||||
15387928
|
Fisherbrand™
88860024 |
1 | 200/240 V, 50/60 Hz (CN, EU, UK) |
£502.00
Each |
In Stock
Log in to see stock. |
|||||
15397928
|
Fisherbrand™
88860025 |
2 | 200/240 V, 50/60 Hz (CN, EU, UK) |
£539.00
Each |
In Stock
Log in to see stock. |
|||||
15317938
|
Fisherbrand™
88860026 |
4 | 200/240 V, 50/60 Hz (CN, EU, UK) |
£615.00
Each |
In Stock
Log in to see stock. |
|||||
Description
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.
Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
- Digital controls and display of time and temperature
- Advanced internal temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
- Powder-coated steel body construction ensures durability
- Built-in over-temperature protection device warrants sample and user safety
- Temperature deviation adjustment
Applications:
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation
Services
2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.
Specifications
CE, cULus, RoHS | |
Powder-coated Steel | |
Ambient +5° to 130°C | |
≤ 20 min. 30° to 130°C | |
≤ ± 1°C | |
2 Year, Replacement | |
3.9 in. | |
50/60 Hz |
PID | |
<80% | |
Dry Bath/Block | |
≤ ± 0.5°C | |
0 to 99:59 min or continuous | |
7.9 in. | |
100 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.