missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Brand: Fisherbrand™ 80863648
Description
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Accessories Offered:
- Well plate blocks with lid
- PCR plate blocks with lid
- Tube blocks
- Conical tube blocks
- Microplate thermal blocks
Specifications
3.9 in. | |
1.2 in. | |
Conical tube block for 50mL tubes | |
Shake Touch Shakers |
9.9 cm | |
3 cm | |
Heating/shaking | |
4 |