Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Brand:  Fisherbrand™ 80863642
 View more versions of this product
Product Code. 15569034
  • £740.00 / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Accessories Offered:
  • Well plate blocks with lid
  • PCR plate blocks with lid
  • Tube blocks
  • Conical tube blocks
  • Microplate thermal blocks
Specifications
Specifications
0.32 in.
0.16 in.
4.7 x 6.4 x 3.0 in. (11.9 x 16.3 x 7.6 cm)
Heating/Shaking
384
0.81 cm
0.4 cm
384-well Plate Block with Lid
Shake Touch Shakers