missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Brand: Fisherbrand™ 80863687
Description
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Accessories Offered:
- Well plate blocks with lid
- PCR plate blocks with lid
- Tube blocks
- Conical tube blocks
- Microplate thermal blocks
Specifications
5 mL, 7 mL | |
1.35 in. | |
0.47 in. | |
4.0 x 5.6 x 2.2 in. (10.2 x 14.2 x 5.6 cm) | |
Heating/shaking | |
24 |
Shaker Accessory Block | |
3.43 cm | |
1.2 cm | |
Tube block for 5 to 7mL tubes | |
Shake Touch Shakers |