missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Label Maker

Easy-Peel™ perforation allows the label backing to be removed quickly and efficiently

Brand:  Fisherbrand™ 1600

Product Code. 11714376

This item is not returnable. View return policy

Description

Description

Quickly and easily make labels for chemicals, bottles, shelves and equipment using this Fisher Scientific™ Lab Label Maker. Easy-Peel™ perforation allows the label backing to be removed quickly and efficiently.

  • Self-adhesive colored tape
  • Sticks powerfully to any surface including glass, plastic, metal, rubber, or wood
  • Raised white letters on 0.375 in. (9mm) wide tape
  • Readable from 8 ft.
  • Easy to operate: Dial a letter, squeeze the handle and pull a lever to cut the tape
Specifications

Specifications

Lab Label Maker
Chemicals, bottles, shelves and equipment
0.05 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Lab Label Maker > Lab Label Maker

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.