missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Long-Stem Digital Thermometer

Durable and safe, containing no mercury or alcohol. Fisherbrand Long-Stem Digital Thermometers are rugged and versatile digital thermometers to read soil, or stir and measure the temperature of solutions. Can also be used with incubators.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.04680kg

Product Code. 11799705

  • £63.42 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Accuracy: ±1°C
  • Large, bright 0.2 in. (6mm) display
  • Reads melting/freezing point temperatures, heat of fusion, temperatures hotter than 110°C (230°F)
  • Stainless-steel probe is resistant to acids, bases and solvents
  • Dual temperature range: -50° to +150°C (-58° to +302°F)
  • Stem length: 8 in. (20.3cm); diameter: 3mm
  • Overall length: 11 in. (28cm)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Specifications

ISO 17025, A2LA, NIST
Food, gases, samples, reagents, water baths and semi solids
8 in.
20 cm
0.1°C
150°C
-50°C to +150°C
Digital LCD
0.14 in.
Stainless Steel probe traceable certificate, protective sleeve, battery
280 mm
ABS Plastic
11 in.
-50°C
±1°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers