Learn More
Fisherbrand™ Maxima C Plus Centering Rings
Centering Ring for use with Maxima C vacuum pumps to join two flanges
Brand: Fisherbrand™ 701073
This item is not returnable.
View return policy
Description
For use with Model M2C and M4C vacuum pumpSpecifications
Model M8C, M12C and M16C vacuum pump | |
Stainless Steel | |
Centering Ring |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Fisherbrand™ Maxima C Plus Centering Rings > Size: NW 25; Fit Models: M8C, M12C, and M16C
Spot an opportunity for improvement?Share a Content Correction