missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ MultiTube Vortexers: Centrifuge Tube Racks
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
Brand: Fisherbrand™ 80862904
This item is not returnable.
View return policy
Description
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
For use with Fisher Scientific MultiTube Vortexers
Color-coded foam racks for centrifuge tubes
*Color subject to change
Specifications
15 mL | |
Centrifuge Tube Rack | |
Multitube vortexer |
15 mL | |
Pink | |
50 |