Learn More
Fisherbrand™ Stand for Model 505 and 705 Sonic Dismembrator
Stand and Converter clamp for Model 505/705 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Brand:  Fisherbrand™ FB459
Product Code. 12883563
  • £192.00 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This Fisher Scientific™ Stand is designed for use with Model 505 and Model 705 Sonic Dismembrators
  • Laboratory Support Stand with Converter Clamp
  • For use with Model 505/705 Sonic Dismembrator
Specifications
Specifications
Stand
61 cm
0.5 in. dia. support rod and converter clamp
24 in.
Model 505 and 705 sonic dismembrator