Learn More
Traceable™ One-Shot™ pH Buffer Standards pH Certified Reference Material (CRM), Fisherbrand™
Performs accurate, reproducible pH calibrations with color-coded bottles. Traceable™ Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
Brand: Fisherbrand
Code : NEW
Additional Details : CAS Number : 7558-79-4 Weight : 0.20000kg
Description
Performs accurate, reproducible pH calibrations with color-coded bottles. Traceable™ Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
- Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Supplied with individual temperature chart, traceability information and Traceable™ Certificate
Specifications
Buffer Standards One-Shot pH 7.000 (CRM) | |
pH Meter and Electrode | |
6 x 100 mL | |
100 mL One-Shot (6 Pk.) |
Yellow | |
7.00 | |
Buffer | |
Certified and Traceable™ |