missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™

Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers.Compatible with all makes of pH instruments and electrodes.

Brand:  Fisherbrand

Code : NEW

Additional Details : CAS Number : 7558-79-4 Weight : 0.50000kg

 View more versions of this product

Product Code. 15508703

  • £34.00 / 500mL
In Stock
to see stock.

Description

Description

Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.

  • Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
  • 100% compatible with all makes of pH instruments and electrodes
  • Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
  • Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
  • Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
  • Supplied withindividual temperature chart, traceability information and Traceable™ Certificate
Specifications

Specifications

Buffer Standards pH 7.000(CRM)
pH Instruments and Electrodes
473 mL
473mL (16 oz.) bottle (ea.)
Yellow
7.000
Buffer
'Certified and Traceable™' ‘ISO 17034 (formerly ISO Guide 34)’ and ‘Certified Reference Material’
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers