missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Excursion-Trac™ Datalogging Thermometer

Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.

Brand:  Fisherbrand™ 90009-95

 View more versions of this product

Product Code. 15204026

  • £151.00 / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Stand, Velcro™, magnetic strips, wall mount, AC power adapter, batteries, and an ISO 17025 Traceable™ Certificate

Description

Description

Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.

High-accuracy thermometer has a temperature range of -50 to 70°C and resolution of 0.01°C and maintains an accuracy of ±0.25°C across the entire temperature range.

  • Stores up to ten unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Datalogging Capacity: 525,600 temperature observations
  • Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
  • New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
Data-Logging Features
  • Monitor readings overnight, on weekends, or for any time period with rolling data log
  • User-defined data-logging interval of one observation/minute to one observation/24 hours
  • Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals
Bottle Probe Features (Cat. Nos. 15-081-125, -126, -128, 129 do not come with bottle probes)
  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples

Note: Units are designed to be used with either the provided AC adaptor or with batteries only. Batteries are designed to last for up to six months. If using battery-power only, the AC power adaptor must be plugged in to download any data.

Order Info

Replacement battery: Cat. No. 1111

Certifications

Supplied with an ISO 17025 Traceable™ Certificate

Specifications

Specifications

Datalogger
525,600 Readings
LED Indicator, Audible, Flashing LCD Segment
Stand, Velcro magnetic strips, wall mount, batteries
USB
±0.25°C
0.01°
-50.00°C to +70.00°C
LCD
Calibrated
2 Bottle Probes
ISO 17025, A2LA, NIST
Temperature-Buffered Bottle Sensor
0.01°
-58° to +158°F
10 ft. (Cable)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers