missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Flip-Stick™ Thermometer
Multipurpose, fold-out design makes taking probe readings in any position a snap
£66.98 - £117.65
Specifications
Certifications/Compliance | ISO 17025, A2LA, NIST |
---|---|
Dimensions (L x W x H) | 11 x 0.6 in. (28 x 1.5 cm) |
For Use With (Application) | Food, gases, samples, reagents, water baths and semi solids |
Includes | Stainless Steel probe |
Length (English) Probe | 4.5 in. |
Product Code | Brand | Accuracy | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Price | Quantity & Availability | |||||
11709735
|
Fisherbrand™
|
±0.3°C |
£117.65
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11719735
|
Fisherbrand™
|
±1°C |
£66.98
Each |
Estimated Shipment: 18-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Fisher Scientific™ Traceable™ Flip-Stick™ Thermometer with fold-out design makes taking probe readings in any position a snap.
- Features °F/°C switch
- Stainless-steel probe
- Shockproof and drop-proof up to 5 ft.
- Max/min temperature reading over any period
- Wide temperature range cover most routine measurements
- Position probe at any angle
- Protect probe in fold-up position
- 0.75 in. Display is easy to read
- Rapid reading update—every second
- HOLD switch freezes display to capture a temperature
Certifications
Individually serial-numbered Traceable Certificate is provided from ISO 17025 calibration laboratory accredited by A2LA indicating traceability to standards provided by NIST (National Institute of Standards and Technology).
Specifications
ISO 17025, A2LA, NIST | |
Food, gases, samples, reagents, water baths and semi solids | |
4.5 in. | |
11.4 cm | |
2.4 oz. | |
0.6 in. | |
572°F | |
-58°F | |
-58°F to +572°F | |
1.5 cm |
11 x 0.6 in. (28 x 1.5 cm) | |
Stainless Steel probe | |
280 mm | |
0.1°C | |
68 g | |
11 in. | |
300°C | |
-50°C | |
-50°C to +300°C |