Learn More
Fisherbrand™ Traceable™ Hi-Accuracy Thermometer
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.01000kg
Description
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
- Available with one probe or two probes
- Monitor inlet and outlet simultaneously
- Visual and audible, high and low settable alarms
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
ISO 17025, A2LA, NIST | |
3.05 mm | |
Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater | |
10.79 cm | |
60 in. | |
226 mm | |
15.24 cm | |
5 oz. | |
2.75 in. | |
9 in. | |
70°C | |
-50°C | |
-50°C to +70°C | |
±0.3°C | |
6.98 cm |
0.12 in. | |
9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm) | |
4.25 in. | |
Stainless Steel Probe, Traceable Certificate and Two AA Batteries | |
6.25 in. | |
152.4 cm | |
0.01°C | |
141.7 g | |
Traceable Thermometer | |
158°F | |
-58°F | |
-58°F to +158°F | |
AA | |
LED |