missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Humidity Meter

Retains highest and lowest readings recorded over selected period

Brand:  Fisherbrand™ 90029-44

Product Code. 11597443

  • £509.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 3 ft. (0.9m) cable and 9V battery for 100 hours of continuous use.

Description

Description

Fisher Scientific™ Traceable™ Humidity Meter retains highest and lowest readings recorded over selected period. Results appear on 1.12 in. LCD.

  • Measures relative humidity from 10 to 95%, with 0.1% resolution and ±2% (mid-range), ±4% (ends of range) accuracy
  • Reads temperatures from -18° to +93°C with 0.1° resolution and accuracy of ±1°C (from 0° to 40°C) and ±2°C (at ends of range)
  • Response time <10 seconds
  • RS-232-C serial computer output
  • Miniature probe
  • Shockproof ABS plastic case

Certifications

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST™). Calibrated by an ISO™ 17025 calibration laboratory accredited by A2LA™.

Compliance

Meets Federal CLIA Requirement 493.1207.

Specifications

Specifications

Humidity Meter
0.001
-18°C to +93°C
LCD
Dual recall memories
0°F to 199.9°F
7.12 x 2.75 x 1.25 in. (18 x 7 x 3 cm)
3 ft.
18 cm
21.5 cm
7 cm
3 cm
1.67 cm
0.304 kg
3 ft. cable and 9V battery for 100 hours of continuous use
10 to 95%
±2 to ±4%
±1°C
ABS Plastic
Polymer Film
0.1°F
7.12 in.
8.5 in.
91.44 cm
2.75 in.
1.25 in.
0.66 in.
10.75 oz.
Retains highest and lowest readings recorded over selected period
ISO 17025, A2LA, NIST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.