Learn More
Fisherbrand™ Traceable™ Logger-Trac™ LN2 Temperature Datalogger
External sensor designed for -200°C environments in the lab or in transit
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.09720kg
Includes: To assure accuracy an individually serial-numbered Traceable Certificate is provided from an ISO/IEC 17025 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Requires :
15-081-149: USB Cable for Logger-Trac LN2 and RH/Temperature
15-081-150: MaxiThermal Software
Description
The Fisher Scientific™ Traceable™ Logger-Trac™ LN2 Temperature Datalogger is ideal for monitoring cryo-material in transit as well as reagents, clinical samples, frozen samples, pharmaceuticals and food items in any location. External sensor designed for -200°C environments in the lab or in transit.
- Displays Cumulative Alarm duration from user selected MIN/MAX temperature values
- Integrates with MaxiThermal Software for complete data analysis
- Reusable/Re-programmable
- Stainless steel external probe with PTFE cable 4.35 in. L x 0.14 in. dia.
- Storage Capacity: 64,000 readings
- Internal clock accuracy: 2 minutes per month
- Temperature sensor: Wired chip in glass thermistor
- Battery: CR2450 3V Lithium coin cell
- Battery life: one year of constant operating use, 3 years shelf life
- IP66 rating
- Size: 3.5 x 2.0 x 1.1 in.
- Weight: 72.8g (2.6 ounces)
- Alarm set points (internal and external sensors individually)
- Recording interval
- Start delay
- Unit description
- Time zone
- Temperature unit of measure
- Stop key enable
Certifications
CE marked
Specifications
Cryo-Temp Datalogger | |
64,000 Readings | |
Visual | |
3 V Lithium Coin Cell (CR2450) | |
Stainless steel external probe and cable | |
IP 66 | |
Wired Chip Glass Thermistor | |
Internal: ±0.2°C (2 to 10°C); ±2°C otherwise; External: ±0.2°C (0 to 30°C); ±2°C otherwise | |
0.1° | |
0.14 in. | |
20 in. (Cable) | |
51 cm (Cable) | |
2.6 oz. |
LCD | |
Calibrated | |
MaxiThermal | |
One year of constant operating use, 3 years shelf life | |
CE, Traceable Certificate | |
ABS Plastic | |
External sensor designed for -200°C environments in the lab or in transit | |
2 min./month | |
Internal: -29°C to +72°C, External: -200°C to +72°C | |
3.5 mm | |
4.35 in. | |
11 cm | |
72.8 g |
MaxiThermal™ Software is needed to activate these dataloggers