Learn More
Fisherbrand™ Refrigerator/Freezer Plus™ Thermometer
May be used for liquids, air/gas, semisolids, water baths and refrigerators
Brand: Fisherbrand™
Code : 50
Additional Details : Weight : 0.09000kg
Description
Monitors temperatures accurately in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Refigerator/Freezer Plus™ Thermometer includes sensor with 5mm diameter and 19mm length. May be used for liquids, air/gas, semisolids, water baths and refrigerators.
- Minimum and maximum temperature memory
- Triple display simultaneously shows minimum, maximum and current temperatures
- High and low temperature alarm programmable in 1 degree increments
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Specifications
ISO 17025, A2LA, NIST | |
5 mm | |
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators | |
120 in. | |
304.8 cm | |
4 oz. | |
2.5 in. | |
158°F | |
-58°F | |
-58°F to +158°F | |
Monitoring Temperature in Freezer | |
6.35 cm |
0.18 in. | |
83 x 16 x 64 mm | |
Traceable certificate, AAA battery | |
83 mm | |
0.1°C | |
113.39 g | |
3.25 in. | |
70°C | |
-50°C | |
-50°C to +70°C | |
±0.5°C |