missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™

IUPAC formulation

Brand:  Fisherbrand 90012-05

33.56 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 11744256

  • £40.45 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

IUPAC formulation

  • Compatible with all makes and instruments and electrodes
  • Accuracy at 25°C is ±0.010 pH
Specifications

Specifications

pH Buffer Standards
4.005
Buffer
Bottle
Certified
Instruments and Electrodes
16 oz.
Accuracy at 25°C is ±0010pH
Traceable to NIST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.