missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™

IUPAC formulation

Brand:  Fisherbrand 98769-35

34.17 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 11784256

  • £41.90 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

IUPAC formulation

  • Compatible with all makes and instruments and electrodes
  • Accuracy at 25°C is ±0.010 pH
Specifications

Specifications

pH Buffer Standards
7.00
Buffer
6 shots of 100mL
Certified
Instruments and Electrodes
6 x 100 mL
Accuracy at 25°C is ±0010pH
Traceable to NIST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers